Propylester av gallussyre 97.0-103.0% (by UV method) Ph. Eur., ren

Leverandør: PanReac AppliChem
Warning

141962.1211
APPC141962.1211EA 9175 NOK
APPC141962.1211
Propylester av gallussyre 97.0-103.0% (by UV method) Ph. Eur., ren
Propylester av gallussyre
Formel: C₁₀H₁₂O₅
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002196
CAS-nummer: 121-79-9

Bestill nå

Spesifikasjon Testresultater

Assay (UV, dried substance) 97,0 - 103,0%
Sulphated ash Max. 0,1%
Loss on drying (105 °C) Max. 0,5%
Identity Passes test
Appearance of solution Passes test
Chloride Max. 0,01%
Total chlorine Max. 0,02%
Gallic acid (TLC) Passes test
Zinc (Zn) max. 0,0025%
Form Solid

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR