Karbondisulfid ≥99.5% (by GC) for synthesis

Leverandør: PanReac AppliChem
Danger

161244.1611
APPC161244.1611EA 1900 NOK
APPC161244.1611
Karbondisulfid ≥99.5% (by GC) for synthesis
Karbondisulfid
Formel: CS₂
Kokepunkt: 46 °C
Smeltepunkt: –111 °C
Tetthet: 1,264 g/cm³
MDL nummer: MFCD00011321
CAS-nummer: 75-15-0
EINECS: 200-843-6
FN: 1131
ADR: 3 (6.1),I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Minimum Assay (GC) ≤99.5%
Identity (IR) Passes Test
Water (H₂O) ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR