Dimetylsulfoksid ≥99.9% (by GC) USP, BP, Ph. Eur., pharma grade

Leverandør: PanReac AppliChem

191954.0716 191954.1214 191954.1611 191954.1612
APPC191954.0716EA 0 NOK
APPC191954.0716 APPC191954.1214 APPC191954.1611 APPC191954.1612
Dimetylsulfoksid ≥99.9% (by GC) USP, BP, Ph. Eur., pharma grade
Dimetylsulfoksid
Formel: (CH₃)₂SO
Kokepunkt: 189 °C
Tetthet: 1,104 g/cm³ (20 °C)
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Minimum Assay (G.C.) 99.9%
Identity According to Pharmacopoeias Passes Test
Acidity Passes Test
Non-Volatile Matter 0.01%
Absorbance (UV) Passes Test
Related Substances (G.C.) 0.1%
Substances Darkened by KOH Passes Test
Residual Solvents (Ph.Eur/USP) Passes Test
Dimethyl Sulphone (G.C.) 0.03%
Water (H₂O) 0.1%
Cd, Class 1 (ICH Q3D) Max. 0.5 ppm
Pb, Class 1 (ICH Q3D) Max. 0.5 ppm
As, Class 1 (ICH Q3D) Max. 1.5 ppm
Hg, Class 1 (ICH Q3D) Max. 1.5 ppm
Co, Class 2A (ICH Q3D) Max. 5 ppm
V, Class 2A (ICH Q3D) Max. 10 ppm
Ni, Class 2A (ICH Q3D) Max. 20 ppm
Tl, Class 2B (ICH Q3D) Max. 0.8 ppm
Au, Class 2B (ICH Q3D) Max. 10 ppm
Pd, Class 2B (ICH Q3D) Max. 10 ppm
Ir, Class 2B (ICH Q3D) Max. 10 ppm
Os, Class 2B (ICH Q3D) Max. 10 ppm
Rh, Class 2B (ICH Q3D) Max. 10 ppm
Ru, Class 2B (ICH Q3D) Max. 10 ppm
Se, Class 2B (ICH Q3D) Max. 15 ppm
Ag, Class 2B (ICH Q3D) Max. 15 ppm
Pt, Class 2B (ICH Q3D) Max. 10 ppm
Li, Class 3 (ICH Q3D) Max. 55 ppm
Sb, Class 3 (ICH Q3D) Max. 120 ppm
Ba, Class 3 (ICH Q3D) Max. 140 ppm
Mo, Class 3 (ICH Q3D) Max. 25 ppm
Cu, Class 3 (ICH Q3D) Max. 250 ppm
Sn, Class 3 (ICH Q3D) Max. 600 ppm
Cr, Class 3 (ICH Q3D) Max. 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR