Vaseline soft, lubricant for glass joints, technical grade
Leverandør: PanReac AppliChem
211757.1211
APPC211757.1211EA 1850 NOK
APPC211757.1211
Vaseline soft, lubricant for glass joints, technical grade
Smøremidler
It is a purified mixture of semisolid saturated hydrocarbons having the general formula CnH2n+2 and is obtained from petroleum.
  • White petrolatum
  • Insoluble in water
  • White amorphous mass
Sertifiseringer: Meets BP and USP standards.