Jodmonoklorid solution 0.1 mol/l (0.2N) acc. Wijs for volumetric analysis

Leverandør: PanReac AppliChem

Synonymer: Iodochloride

281590.1611
APPC281590.1611EA 2875 NOK
APPC281590.1611
Jodmonoklorid solution 0.1 mol/l (0.2N) acc. Wijs for volumetric analysis
Jodmonoklorid
Formel: ClI
MDL nummer: MFCD00011354
CAS-nummer: 7790-99-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR