Oxidase sticks for microbiology
Leverandør: PanReac AppliChem
416444.2326
APPC416444.2326EA 1800 NOK
APPC416444.2326
Oxidase sticks for microbiology
Mikrobiologiske testsystemer Enzymatiske tester
Plastic sticks used for the easy and rapid detection of the cytochrome oxidase enzyme in microbiological diagnosis. Oxidase sticks have a much higher stability compared to the traditional laboratory test, because the tetramethyl-p-phenylenediamine reagent is attached to the pad.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904