Contact plate, violet red bile glucose (VRBG) agar

Leverandør: PanReac AppliChem

433745.0922
APPC433745.0922EA 2175 NOK
APPC433745.0922
Contact plate, violet red bile glucose (VRBG) agar
Mikrobiologiske medier Kontaktplater

Violet red bile glucose agar (VRBG) is used as a culture media for microbiology.


  • pH: 7,4 ±0,2
  • Incubate at 30 to 35 °C for 24 to 48 hours
  • Storage in a cool and dry place


It contains bile salts mixture (1,5 g/L), crystal violet (0,002 g/L), neutral red (0,03 g/L), D(+)-glucose (10,0 g/L), yeast extract (3,0 g/L), gelatin peptone (7,0 g/L), sodium chloride (5,0 g/L) and agar (20,5 g/L).


Sertifiseringer: Meets Ph. Eur. and ISO 21528 standard specifications.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR