N,N-Dimetylacetamid ≥99.9% (by GC) for headspace gas chromatography

Leverandør: PanReac AppliChem

753145.1611
APPC753145.1611EA 5900 NOK
APPC753145.1611
N,N-Dimetylacetamid ≥99.9% (by GC) for headspace gas chromatography
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
Smeltepunkt: –20 °C
Flash punkt: 70 °C
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Minimum Assay (G.C.) 99,9 %
Identity IR Passes Test
Refractive Index n20/D 1.438 - 1.439
Apha Colour ≤10
Suitability for Analysis of Residual Solvents Cg/Hs ≤0.5 ppm
Solvents of Class 1 According To ICH ≤5 ppm
Solvents of Class 2 According To ICH ≤25 ppm
Solvents Of Class 3 According To ICH ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR