D(-)-Mannitol

Leverandør: PanReac AppliChem

A4831.1000
APPCA4831.1000EA 10450 NOK
APPCA4831.1000
D(-)-Mannitol
D(-)-Mannitol
Formel: HOH₂C(CH(OH))₄CH₂OH
Kokepunkt: 290…295 °C (4 hPa)
Smeltepunkt: 164…169 °C
Tetthet: 1,49 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064287
CAS-nummer: 69-65-8
EINECS: 200-711-8
Merck Indeks: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR