Folin-Ciocalteu's reagent Phenols_analysis

Leverandør: PanReac AppliChem
Warning

Synonymer: Folin & Ciocalteu’s phenol reagent

A5084.0500
APPCA5084.0500EA 3400 NOK
APPCA5084.0500
Folin-Ciocalteu's reagent Phenols_analysis
Folin-Ciocalteu's reagent
Tetthet: 1.22 g/cm³ (20 °C)
MDL nummer: MFCD00132625
FN: 3264
ADR: 8 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR