DrySyn® MAXI Heating Blocks, 2 L
Leverandør: ASYNT
ADS2-2
ASYNADS2-2EA 0 NOK
ASYNADS2-2
DrySyn® MAXI Heating Blocks, 2 L
Termostatblokker Tilbehør til termostatblokker
The DrySyn® MAXI 2 L is a stand-alone 2000 ml version of the standard DrySyn MAXI Kit (which alternatively has a 3000 ml base and 2000 ml insert) which comes with heat resistant safety lifting handles.
  • Enables clean, safe synthesis without the hazards or problems associated with silicon oil baths or heating mantles
  • With no messy oil, heat resistant handles, and simple to configure range of inserts this system is world renowned
  • With heating rates the same as an oil bath there is no need to change your chemistry either
  • Fits standard hotplate stirrers with