Anti-GAPDHS Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP40187_P050
AVIVARP40187_P050EA 4400 NOK
AVIVARP40187_P050
Anti-GAPDHS Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GAPDHS (glyceraldehyde-3-phosphate dehydrogenase, spermatogenic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol GAPDHS
Antigen-navn glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
Environmentally Preferable
Antigen synonymer GAPDH-2|HSD-35|GAPD2|GAPDS
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 26330
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR