Anti-EIF2A Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP41041_T100
AVIVARP41041_T100EA 3850 NOK
AVIVARP41041_T100
Anti-EIF2A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: EIF2A (eukaryotic translation initiation factor 2A, 65kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR