Anti-POLDIP3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP41048_P050
AVIVARP41048_P050EA 4400 NOK
AVIVARP41048_P050
Anti-POLDIP3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: POLDIP3 (polymerase (DNA-directed), delta interacting protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol POLDIP3
Antigen-navn Polymerase (DNA-directed), delta interacting protein 3
Environmentally Preferable
Antigen synonymer PDIP46|SKAR
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 84271
Isotype
Western blot Yes
Epitop C-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR