Anti-NXF5 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP41062_T100
AVIVARP41062_T100EA 3850 NOK
AVIVARP41062_T100
Anti-NXF5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NXF5 (nuclear RNA export factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol NXF5
Antigen-navn nuclear RNA export factor 5
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 55998
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR