Anti-SRSF10 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP41083_P050
AVIVARP41083_P050EA 4400 NOK
AVIVARP41083_P050
Anti-SRSF10 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SRSF10 (serine/arginine-rich splicing factor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR