Anti-A1CF Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP41109_P050
AVIVARP41109_P050EA 4400 NOK
AVIVARP41109_P050
Anti-A1CF Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: A1CF (APOBEC1 complementation factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR