Anti-SLC6A8 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP42248_T100
AVIVARP42248_T100EA 3850 NOK
AVIVARP42248_T100
Anti-SLC6A8 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC6A8 (solute carrier family 6 (neurotransmitter transporter), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR