Anti-ATP6V0A2 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP42365_P050
AVIVARP42365_P050EA 4400 NOK
AVIVARP42365_P050
Anti-ATP6V0A2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ATP6V0A2 (ATPase, H+ transporting, lysosomal V0 subunit a2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR