Anti-SLC41A2 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP44062_T100
AVIVARP44062_T100EA 3850 NOK
AVIVARP44062_T100
Anti-SLC41A2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC41A2 (solute carrier family 41 (magnesium transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR