Anti-SLC46A3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP44158_T100
AVIVARP44158_T100EA 3850 NOK
AVIVARP44158_T100
Anti-SLC46A3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC46A3 (solute carrier family 46, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol SLC46A3
Antigen-navn solute carrier family 46, member 3
Environmentally Preferable
Antigen synonymer FKSG16
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 283537
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR