Anti-SLC19A1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP44167_T100
AVIVARP44167_T100EA 3850 NOK
AVIVARP44167_T100
Anti-SLC19A1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC19A1 (solute carrier family 19 (folate transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR