Anti-SLC25A35 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP44177_P050
AVIVARP44177_P050EA 4400 NOK
AVIVARP44177_P050
Anti-SLC25A35 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC25A35 (solute carrier family 25, member 35)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR