Anti-NAT2 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP44195_T100
AVIVARP44195_T100EA 3850 NOK
AVIVARP44195_T100
Anti-NAT2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NAT2 (N-acetyltransferase 2 (arylamine N-acetyltransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR