Anti-EPOR Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP44217_P050
AVIVARP44217_P050EA 4400 NOK
AVIVARP44217_P050
Anti-EPOR Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: EPOR (erythropoietin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR