Anti-RORA Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP45607_P050
AVIVARP45607_P050EA 4400 NOK
AVIVARP45607_P050
Anti-RORA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RORA (RAR-related orphan receptor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol RORA
Environmentally Preferable
Antigen-navn RAR-related orphan receptor A
Antigen synonymer RZRA|RZR-ALPHA|ROR3|ROR1|NR1F1|ROR2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 6095
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR