Anti-NR1H2 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP45633_T100
AVIVARP45633_T100EA 3850 NOK
AVIVARP45633_T100
Anti-NR1H2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NR1H2 (nuclear receptor subfamily 1, group H, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR