Anti-NR1D1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP45634_P050
AVIVARP45634_P050EA 4400 NOK
AVIVARP45634_P050
Anti-NR1D1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NR1D1 (nuclear receptor subfamily 1, group D, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR