Anti-RXRB Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP45637_P050
AVIVARP45637_P050EA 4400 NOK
AVIVARP45637_P050
Anti-RXRB Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RXRB (retinoid X receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol RXRB
Antigen-navn retinoid X receptor, beta
Environmentally Preferable
Antigen synonymer RCoR-1|H-2RIIBP|DAUDI6|NR2B2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Pig,Human,Bovine,Guinea pig,Dog,Rat,Mouse
Vert Rabbit
Gen-ID 6257
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR