Anti-PTDSS1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47068_P050
AVIVARP47068_P050EA 4400 NOK
AVIVARP47068_P050
Anti-PTDSS1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PTDSS1 (phosphatidylserine synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR