Anti-LRRC8B Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47145_P050
AVIVARP47145_P050EA 4400 NOK
AVIVARP47145_P050
Anti-LRRC8B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LRRC8B (leucine rich repeat containing 8 family, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol LRRC8B
Environmentally Preferable
Antigen-navn leucine rich repeat containing 8 family, member B
Antigen synonymer TALRRP|TA-LRRP
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 23507
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR