Anti-TEX264 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47200_P050
AVIVARP47200_P050EA 4400 NOK
AVIVARP47200_P050
Anti-TEX264 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TEX264 (testis expressed 264)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol TEX264
Environmentally Preferable
Antigen-navn Testis Expressed 264
Antigen synonymer ZSIG11
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 51368
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR