Anti-ERGIC3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47209_P050
AVIVARP47209_P050EA 4400 NOK
AVIVARP47209_P050
Anti-ERGIC3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ERGIC3 (ERGIC and golgi 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol ERGIC3
Antigen-navn ERGIC and golgi 3
Environmentally Preferable
Antigen synonymer PRO0989|Erv46|CGI-54|NY-BR-84|C20orf47|dJ477O4.2|SDBCAG84
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 51614
Isotype
Western blot Yes
Epitop C-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR