Anti-HSD17B12 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47217_P050
AVIVARP47217_P050EA 4400 NOK
AVIVARP47217_P050
Anti-HSD17B12 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSD17B12 (hydroxysteroid (17-beta) dehydrogenase 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR