Anti-SDF4 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47223_P050
AVIVARP47223_P050EA 4400 NOK
AVIVARP47223_P050
Anti-SDF4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SDF4 (stromal cell derived factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol SDF4
Antigen-navn Stromal cell-derived factor 4
Environmentally Preferable
Antigen synonymer Cab45|SDF-4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 51150
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR