Anti-CLEC1B Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47260_P050
AVIVARP47260_P050EA 4400 NOK
AVIVARP47260_P050
Anti-CLEC1B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CLEC1B (C-type lectin domain family 1, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR