Anti-GOLM1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47269_P050
AVIVARP47269_P050EA 4400 NOK
AVIVARP47269_P050
Anti-GOLM1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GOLM1 (golgi membrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol GOLM1
Environmentally Preferable
Antigen-navn Golgi membrane protein 1
Antigen synonymer HEL46|GP73|C9orf155|GOLPH2|bA379P1.3|PSEC0257
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 51280
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR