Anti-TMEM104 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP47331_T100
AVIVARP47331_T100EA 3850 NOK
AVIVARP47331_T100
Anti-TMEM104 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TMEM104 (transmembrane protein 104)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol TMEM104
Environmentally Preferable
Antigen-navn transmembrane protein 104
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 54868
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR