Anti-MAP4K2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP48631_P050
AVIVARP48631_P050EA 4400 NOK
AVIVARP48631_P050
Anti-MAP4K2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAP4K2 (mitogen-activated protein kinase kinase kinase kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR