Anti-SULT2B1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48636_P050
AVIVARP48636_P050EA 4400 NOK
AVIVARP48636_P050
Anti-SULT2B1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SULT2B1 (sulfotransferase family, cytosolic, 2B, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR