Anti-CDY1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48645_T100
AVIVARP48645_T100EA 3850 NOK
AVIVARP48645_T100
Anti-CDY1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CDY1 (chromodomain protein, Y-linked, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR