Anti-OXSR1 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48661_P050
AVIVARP48661_P050EA 4400 NOK
AVIVARP48661_P050
Anti-OXSR1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: OXSR1 (oxidative stress responsive 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol OXSR1
Environmentally Preferable
Antigen-navn oxidative-stress responsive 1
Antigen synonymer 2210022N24Rik|AI462649|AW209236|Osr1|2810422B09Rik|mKIAA1101
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 108737
Isotype
Western blot Yes
Epitop middle
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR