Anti-STK39 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48744_T100
AVIVARP48744_T100EA 3850 NOK
AVIVARP48744_T100
Anti-STK39 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STK39 (serine threonine kinase 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol STK39
Antigen-navn Serine Threonine Kinase 39
Environmentally Preferable
Antigen synonymer PASK|SPAK|DCHT
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 27347
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR