Anti-NUDT13 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48782_P050
AVIVARP48782_P050EA 4400 NOK
AVIVARP48782_P050
Anti-NUDT13 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NUDT13 (nudix (nucleoside diphosphate linked moiety X)-type motif 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR