Anti-POLR1D Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48787_P050
AVIVARP48787_P050EA 4400 NOK
AVIVARP48787_P050
Anti-POLR1D Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: POLR1D (polymerase (RNA) I polypeptide D, 16kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Dog
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR