Anti-TRMU Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48818_P050
AVIVARP48818_P050EA 4400 NOK
AVIVARP48818_P050
Anti-TRMU Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRMU (tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol TRMU
Environmentally Preferable
Antigen-navn tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Antigen synonymer TRMT|TRNT1|MTO2|TRMT1|LCAL3|MTU1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 55687
Isotype
Western blot Yes
Epitop N-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR