Anti-PCMTD2 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP48826_P050
AVIVARP48826_P050EA 4400 NOK
AVIVARP48826_P050
Anti-PCMTD2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PCMTD2 (protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol PCMTD2
Environmentally Preferable
Antigen-navn protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
Antigen synonymer 5330414D10Rik|AI314201
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Gen-ID 245867
Isotype
Western blot Yes
Epitop C-terminal
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR