Anti-HLA-F Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP49410_P050
AVIVARP49410_P050EA 4400 NOK
AVIVARP49410_P050
Anti-HLA-F Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HLA-F (major histocompatibility complex, class I, F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR