Anti-JAM3 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP50121_P050
AVIVARP50121_P050EA 4400 NOK
AVIVARP50121_P050
Anti-JAM3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: JAM3 (junctional adhesion molecule 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR