Anti-ZC2HC1B Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
ARP51126_P050
AVIVARP51126_P050EA 4400 NOK
AVIVARP51126_P050
Anti-ZC2HC1B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZC2HC1B (zinc finger, C2HC-type containing 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR